Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLK13
Entrez Gene ID
26085
Gene Name
kallikrein related peptidase 13
Gene Aliases
KLK-L4, KLKL4
Chromosome Location
Chr.19: 51056206 - 51065110 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015596.1 NP_056411.1 3 - 4 552 125
GenBank mRNA AB451465.1 - 3 - 4 509 125
BC069334.1 - 3 - 4 553 125
BC069543.1 - 3 - 4 552 125

View Full Product Details