Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAP3K14
Entrez Gene ID
9020
Gene Name
mitogen-activated protein kinase kinase kinase 14
Gene Aliases
FTDCR1B, HS, HSNIK, NIK
Chromosome Location
Chr.17: 45263119 - 45317064 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003954.4 NP_003945.2 6 - 7 1417 76
XM_011525441.2 XP_011523743.1 7 - 8 1554 76
GenBank mRNA AK290203.1 - 6 - 7 1393 76
AK313085.1 - 6 - 7 1393 76
BC035576.1 - 6 - 7 1362 76
CR749592.1 - 3 - 4 641 76
Y10256.1 - 5 - 6 1524 76

View Full Product Details