The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020307.3 | NP_064703.1 | 10 - 11 | 1897 | 95 |
XM_005247648.3 | XP_005247705.1 | 11 - 12 | 1354 | 95 | |
XM_011513011.2 | XP_011511313.1 | 11 - 12 | 1906 | 95 | |
XM_017006891.1 | XP_016862380.1 | 11 - 12 | 1460 | 95 | |
XM_017006892.1 | XP_016862381.1 | 11 - 12 | 3026 | 95 | |
XM_017006893.1 | XP_016862382.1 | 11 - 12 | 2934 | 95 | |
XM_017006894.1 | XP_016862383.1 | 12 - 13 | 1491 | 95 | |
XM_017006895.1 | XP_016862384.1 | 12 - 13 | 1368 | 95 | |
XM_017006896.1 | XP_016862385.1 | 11 - 12 | 1295 | 95 | |
GenBank mRNA | AF180920.1 | - | 10 - 11 | 1285 | 95 |
AF208843.1 | - | 6 - 7 | 718 | 95 | |
AF367476.1 | - | 12 - 13 | 1539 | 95 | |
AK022974.1 | - | 10 - 11 | 1284 | 95 | |
AY358663.1 | - | 5 - 6 | 894 | 95 | |
BC007081.1 | - | 10 - 11 | 1284 | 95 | |
BC038394.1 | - | 12 - 13 | 1547 | 95 | |
BC067812.1 | - | 10 - 11 | 1273 | 95 | |
BX641146.1 | - | 5 - 6 | 4004 | 95 |