The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031231.3 | NP_112508.3 | 12 - 13 | 1159 | 100 |
NM_031232.3 | NP_112509.3 | 11 - 12 | 1261 | 100 | |
XM_005260510.1 | XP_005260567.1 | 11 - 12 | 1075 | 100 | |
XM_011528991.1 | XP_011527293.1 | 8 - 9 | 785 | 100 | |
XM_017028015.1 | XP_016883504.1 | 8 - 9 | 784 | 100 | |
XM_017028016.1 | XP_016883505.1 | 8 - 9 | 713 | 100 | |
GenBank mRNA | AB039947.1 | - | 9 - 10 | 963 | 100 |
AF409141.1 | - | 12 - 13 | 1107 | 100 | |
AK023706.1 | - | 11 - 12 | 1365 | 100 | |
AK124388.1 | - | 8 - 9 | 1233 | 100 | |
AK291895.1 | - | 12 - 13 | 1089 | 100 | |
BC047673.1 | - | 12 - 13 | 1114 | 100 | |
CD580591.2 | - | 1 - 2 | 99 | 100 |