The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003810.1 | NP_001003810.1 | 6 - 7 | 1215 | 114 |
NM_002138.3 | NP_002129.2 | 7 - 8 | 1272 | 114 | |
NM_031369.2 | NP_112737.1 | 7 - 8 | 1362 | 114 | |
NM_031370.2 | NP_112738.1 | 8 - 9 | 1419 | 114 | |
GenBank mRNA | AF039575.1 | - | 7 - 8 | 930 | 114 |
AK057836.1 | - | 8 - 9 | 1380 | 114 | |
AK300149.1 | - | 8 - 9 | 1251 | 114 | |
BC002401.1 | - | 8 - 9 | 1165 | 114 | |
BC023977.2 | - | 7 - 8 | 1247 | 114 | |
BC026015.2 | - | 7 - 8 | 1247 | 114 | |
D55671.1 | - | 7 - 8 | 838 | 114 | |
D55672.1 | - | 7 - 8 | 1239 | 114 | |
D55674.1 | - | 8 - 9 | 1365 | 114 |