The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003800.1 | NP_001003800.1 | 2 - 3 | 516 | 66 |
NM_015250.3 | NP_056065.1 | 2 - 3 | 516 | 66 | |
XM_017014551.1 | XP_016870040.1 | 2 - 3 | 637 | 66 | |
GenBank mRNA | AB014599.1 | - | 2 - 3 | 436 | 66 |
AY052562.1 | - | 2 - 3 | 516 | 66 | |
BC073970.1 | - | 2 - 3 | 531 | 66 |