Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCTN4
Entrez Gene ID
51164
Gene Name
dynactin subunit 4
Gene Aliases
DYN4, P62
Chromosome Location
Chr.5: 150708747 - 150759095 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135643.1 NP_001129115.1 2 - 3 312 64
NM_001135644.1 NP_001129116.1 2 - 3 480 64
NM_016221.3 NP_057305.1 2 - 3 312 64
XM_005268457.1 XP_005268514.1 2 - 3 250 64
XM_011537644.1 XP_011535946.1 3 - 4 216 64
XM_011537645.1 XP_011535947.1 3 - 4 128 64
GenBank mRNA AF195120.1 - 2 - 3 227 64
AK000299.1 - 2 - 3 231 64
AK125973.1 - 2 - 3 480 64
AK296787.1 - 2 - 3 231 64
AK299517.1 - 2 - 3 355 64
AK307757.1 - 2 - 3 251 64
AK311693.1 - 2 - 3 248 64
BC026323.1 - 2 - 3 237 64
DC321382.1 - 2 - 3 312 64
KU178637.1 - 2 - 3 210 64
KU178638.1 - 2 - 3 210 64

View Full Product Details