The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032451.1 | NP_115827.1 | 11 - 12 | 1763 | 74 |
XM_011523379.1 | XP_011521681.1 | 11 - 12 | 2529 | 74 | |
XM_011523380.2 | XP_011521682.1 | 12 - 13 | 1785 | 74 | |
XM_011523381.1 | XP_011521683.1 | 12 - 13 | 2609 | 74 | |
XM_011523384.1 | XP_011521686.1 | 11 - 12 | 1787 | 74 | |
XM_017023779.1 | XP_016879268.1 | 11 - 12 | 1775 | 74 | |
XM_017023780.1 | XP_016879269.1 | 11 - 12 | 1940 | 74 | |
XM_017023781.1 | XP_016879270.1 | 10 - 11 | 1412 | 74 | |
GenBank mRNA | AB058735.1 | - | 9 - 10 | 1861 | 74 |
AJ422077.1 | - | 11 - 12 | 1753 | 74 | |
AK094918.1 | - | 4 - 5 | 580 | 74 | |
AK130685.1 | - | 1 - 2 | 1645 | 74 | |
AL834408.1 | - | 11 - 12 | 1557 | 74 | |
BC063706.1 | - | 3 - 4 | 376 | 74 | |
BC111030.1 | - | 11 - 12 | 1763 | 74 | |
BC139732.1 | - | 9 - 10 | 1381 | 74 |