Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPIRE2
Entrez Gene ID
84501
Gene Name
spire type actin nucleation factor 2
Gene Aliases
Spir-2
Chromosome Location
Chr.16: 89817477 - 89871319 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032451.1 NP_115827.1 2 - 3 342 76
XM_011523379.1 XP_011521681.1 2 - 3 1108 76
XM_011523384.1 XP_011521686.1 2 - 3 366 76
XM_011523385.1 XP_011521687.1 2 - 3 366 76
XM_017023779.1 XP_016879268.1 2 - 3 354 76
XM_017023780.1 XP_016879269.1 2 - 3 519 76
GenBank mRNA AJ422077.1 - 2 - 3 332 76
AL834408.1 - 2 - 3 136 76
BC111030.1 - 2 - 3 342 76

View Full Product Details