The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020971.2 | NP_066022.2 | 11 - 12 | 1469 | 64 |
XM_011527173.2 | XP_011525475.1 | 11 - 12 | 1535 | 64 | |
XM_017027049.1 | XP_016882538.1 | 11 - 12 | 1722 | 64 | |
XM_017027050.1 | XP_016882539.1 | 9 - 10 | 1104 | 64 | |
XM_017027051.1 | XP_016882540.1 | 11 - 12 | 1535 | 64 | |
XM_017027052.1 | XP_016882541.1 | 11 - 12 | 1535 | 64 | |
GenBank mRNA | AB046862.1 | - | 2 - 3 | 212 | 64 |
AF082075.1 | - | 11 - 12 | 1545 | 64 | |
AF311855.1 | - | 11 - 12 | 1469 | 64 | |
AF324063.1 | - | 11 - 12 | 1453 | 64 | |
AF324064.1 | - | 11 - 12 | 1453 | 64 | |
AY004226.1 | - | 10 - 11 | 1367 | 64 | |
AY004227.1 | - | 10 - 11 | 1367 | 64 |