Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PTH2
Entrez Gene ID
113091
Gene Name
parathyroid hormone 2
Gene Aliases
TIP39
Chromosome Location
Chr.19: 49422414 - 49423441 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178449.3 NP_848544.1 1 - 2 230 156
GenBank mRNA AY037555.1 - 1 - 2 230 156
AY048588.1 - 1 - 2 131 156
BC093995.1 - 1 - 2 169 156
BC093997.1 - 1 - 2 173 156

View Full Product Details