The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001287529.1 | NP_001274458.1 | 5 - 6 | 1082 | 81 |
NM_174983.4 | NP_778148.2 | 5 - 6 | 1109 | 81 | |
XM_005259490.4 | XP_005259547.1 | 5 - 6 | 1117 | 81 | |
XM_006722647.3 | XP_006722710.1 | 5 - 6 | 1116 | 81 | |
XM_011527684.2 | XP_011525986.1 | 5 - 6 | 1115 | 81 | |
XM_017026288.1 | XP_016881777.1 | 5 - 6 | 1116 | 81 | |
GenBank mRNA | BC036706.2 | - | 5 - 6 | 1061 | 81 |
BC068439.1 | - | 5 - 6 | 1098 | 81 |