The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300959.1 | NP_001287888.1 | 14 - 15 | 1945 | 73 |
NM_031449.3 | NP_113637.3 | 15 - 16 | 2114 | 73 | |
NM_174929.2 | NP_777589.2 | 13 - 14 | 1975 | 73 | |
XM_005249866.2 | XP_005249923.1 | 15 - 16 | 2165 | 73 | |
XM_005249867.4 | XP_005249924.1 | 15 - 16 | 2167 | 73 | |
XM_005249868.4 | XP_005249925.1 | 15 - 16 | 3185 | 73 | |
XM_005249869.2 | XP_005249926.1 | 15 - 16 | 2138 | 73 | |
XM_005249870.2 | XP_005249927.1 | 14 - 15 | 2087 | 73 | |
XM_005249871.2 | XP_005249928.1 | 15 - 16 | 2069 | 73 | |
XM_005249872.2 | XP_005249929.1 | 14 - 15 | 2060 | 73 | |
XM_005249873.2 | XP_005249930.1 | 15 - 16 | 2042 | 73 | |
XM_011515566.1 | XP_011513868.1 | 9 - 10 | 1305 | 73 | |
XM_017012674.1 | XP_016868163.1 | 15 - 16 | 2246 | 73 | |
GenBank mRNA | AB067473.1 | - | 9 - 10 | 1198 | 73 |
AF357908.1 | - | 14 - 15 | 1997 | 73 | |
AK090415.1 | - | 15 - 16 | 2114 | 73 | |
AK090841.1 | - | 7 - 8 | 1394 | 73 | |
AL136572.1 | - | 5 - 6 | 567 | 73 | |
AL831933.1 | - | 6 - 7 | 1712 | 73 | |
AY426594.1 | - | 13 - 14 | 1937 | 73 | |
BC016968.1 | - | 13 - 14 | 1958 | 73 | |
BC021924.1 | - | 9 - 10 | 1046 | 73 | |
BC110435.1 | - | 14 - 15 | 1930 | 73 |