The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300959.1 | NP_001287888.1 | 4 - 5 | 401 | 111 |
NM_031449.3 | NP_113637.3 | 4 - 5 | 492 | 111 | |
NM_174929.2 | NP_777589.2 | 3 - 4 | 431 | 111 | |
XM_005249866.2 | XP_005249923.1 | 4 - 5 | 516 | 111 | |
XM_005249867.4 | XP_005249924.1 | 4 - 5 | 518 | 111 | |
XM_005249868.4 | XP_005249925.1 | 4 - 5 | 1536 | 111 | |
XM_005249869.2 | XP_005249926.1 | 4 - 5 | 516 | 111 | |
XM_005249870.2 | XP_005249927.1 | 4 - 5 | 516 | 111 | |
XM_005249871.2 | XP_005249928.1 | 4 - 5 | 420 | 111 | |
XM_005249872.2 | XP_005249929.1 | 4 - 5 | 516 | 111 | |
XM_005249873.2 | XP_005249930.1 | 4 - 5 | 420 | 111 | |
XM_017012674.1 | XP_016868163.1 | 4 - 5 | 597 | 111 | |
GenBank mRNA | AF357908.1 | - | 3 - 4 | 369 | 111 |
AK090415.1 | - | 4 - 5 | 492 | 111 | |
AY426594.1 | - | 3 - 4 | 108 | ||
BC016968.1 | - | 3 - 4 | 414 | 111 | |
BC110435.1 | - | 4 - 5 | 386 | 111 |