The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000946.2 | NP_000937.1 | 5 - 6 | 644 | 91 |
GenBank mRNA | AK223391.1 | - | 5 - 6 | 605 | 91 |
AK314866.1 | - | 5 - 6 | 605 | 91 | |
BC005266.1 | - | 5 - 6 | 617 | 91 | |
BU167590.1 | - | 5 - 6 | 644 | 91 | |
X74330.1 | - | 5 - 6 | 605 | 91 |