The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286561.1 | NP_001273490.1 | 11 - 12 | 1174 | 107 |
NM_006185.3 | NP_006176.2 | 10 - 11 | 1076 | 107 | |
NR_104476.1 | 10 - 11 | 995 | 107 | ||
XM_006718564.1 | XP_006718627.1 | 9 - 10 | 891 | 107 | |
XM_011545054.1 | XP_011543356.1 | 9 - 10 | 891 | 107 | |
XM_011545055.1 | XP_011543357.1 | 10 - 11 | 1069 | 107 | |
XM_011545056.2 | XP_011543358.1 | 11 - 12 | 1063 | 107 | |
XM_011545057.2 | XP_011543359.1 | 11 - 12 | 1053 | 107 | |
XM_011545058.1 | XP_011543360.1 | 9 - 10 | 1090 | 107 | |
XM_011545059.2 | XP_011543361.1 | 11 - 12 | 1055 | 107 | |
XM_011545060.2 | XP_011543362.1 | 11 - 12 | 1049 | 107 | |
XM_011545061.2 | XP_011543363.1 | 9 - 10 | 909 | 107 | |
XM_011545062.2 | XP_011543364.1 | 9 - 10 | 922 | 107 | |
XM_011545063.2 | XP_011543365.1 | 9 - 10 | 889 | 107 | |
XM_011545064.1 | XP_011543366.1 | 9 - 10 | 876 | 107 | |
XM_011545065.1 | XP_011543367.1 | 9 - 10 | 891 | 107 | |
XM_011545066.1 | XP_011543368.1 | 9 - 10 | 891 | 107 | |
XM_017017830.1 | XP_016873319.1 | 10 - 11 | 931 | 107 | |
XM_017017831.1 | XP_016873320.1 | 9 - 10 | 927 | 107 | |
GenBank mRNA | AB210007.1 | - | 10 - 11 | 877 | 107 |
AK290509.1 | - | 9 - 10 | 891 | 107 | |
AK307577.1 | - | 10 - 11 | 894 | 107 | |
AK309041.1 | - | 9 - 10 | 898 | 107 | |
BC004165.2 | - | 10 - 11 | 910 | 107 | |
BC027493.1 | - | 10 - 11 | 877 | 107 | |
BC043499.1 | - | 10 - 11 | 877 | 107 | |
BC103765.1 | - | 10 - 11 | 995 | 107 | |
Z11583.1 | - | 10 - 11 | 894 | 107 | |
Z11584.1 | - | 11 - 12 | 996 | 107 |