The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286561.1 | NP_001273490.1 | 23 - 24 | 6223 | 83 |
NM_006185.3 | NP_006176.2 | 23 - 24 | 6167 | 83 | |
NR_104476.1 | 21 - 22 | 2678 | 83 | ||
XM_006718564.1 | XP_006718627.1 | 22 - 23 | 5982 | 83 | |
XM_011545054.1 | XP_011543356.1 | 23 - 24 | 6036 | 83 | |
XM_011545055.1 | XP_011543357.1 | 24 - 25 | 6214 | 83 | |
XM_011545056.2 | XP_011543358.1 | 25 - 26 | 6208 | 83 | |
XM_011545057.2 | XP_011543359.1 | 25 - 26 | 6198 | 83 | |
XM_011545058.1 | XP_011543360.1 | 23 - 24 | 6235 | 83 | |
XM_011545059.2 | XP_011543361.1 | 25 - 26 | 6200 | 83 | |
XM_011545060.2 | XP_011543362.1 | 25 - 26 | 6194 | 83 | |
XM_011545061.2 | XP_011543363.1 | 23 - 24 | 6054 | 83 | |
XM_011545062.2 | XP_011543364.1 | 23 - 24 | 6067 | 83 | |
XM_011545063.2 | XP_011543365.1 | 23 - 24 | 6034 | 83 | |
XM_011545064.1 | XP_011543366.1 | 23 - 24 | 6021 | 83 | |
XM_011545065.1 | XP_011543367.1 | 22 - 23 | 5994 | 83 | |
XM_011545066.1 | XP_011543368.1 | 21 - 22 | 5940 | 83 | |
XM_017017830.1 | XP_016873319.1 | 22 - 23 | 5980 | 83 | |
XM_017017831.1 | XP_016873320.1 | 21 - 22 | 5976 | 83 | |
GenBank mRNA | AB208841.1 | - | 8 - 9 | 4240 | 83 |
AB210007.1 | - | 23 - 24 | 5968 | 83 | |
BC004165.2 | - | 21 - 22 | 2593 | 83 | |
BC008345.1 | - | 9 - 10 | 2375 | 83 | |
BC013023.2 | - | 8 - 9 | 3069 | 83 | |
Z11583.1 | - | 23 - 24 | 5985 | 83 | |
Z11584.1 | - | 23 - 24 | 6045 | 83 | |
Z14229.1 | - | 6 - 7 | 1104 | 83 |