Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CPNE7
Entrez Gene ID
27132
Gene Name
copine 7
Gene Aliases
-
Chromosome Location
Chr.16: 89575768 - 89597246 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014427.4 NP_055242.1 5 - 6 788 60
NM_153636.2 NP_705900.1 3 - 4 563 60
XM_011523000.1 XP_011521302.1 3 - 4 558 60
XM_011523001.2 XP_011521303.1 3 - 4 323 60
XM_017023138.1 XP_016878627.1 5 - 6 752 60
XM_017023139.1 XP_016878628.1 5 - 6 763 60
XM_017023140.1 XP_016878629.1 3 - 4 538 60
XM_017023141.1 XP_016878630.1 2 - 3 1645 60
GenBank mRNA AJ133798.1 - 5 - 6 787 60
AL539371.3 - 3 - 4 564 60
BC035334.1 - 3 - 4 501 60
BC064577.1 - 3 - 4 540 60

View Full Product Details