The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014427.4 | NP_055242.1 | 13 - 14 | 1468 | 88 |
NM_153636.2 | NP_705900.1 | 11 - 12 | 1243 | 88 | |
XM_011523000.1 | XP_011521302.1 | 11 - 12 | 1238 | 88 | |
XM_011523001.2 | XP_011521303.1 | 11 - 12 | 1003 | 88 | |
XM_017023138.1 | XP_016878627.1 | 13 - 14 | 1432 | 88 | |
XM_017023139.1 | XP_016878628.1 | 13 - 14 | 1443 | 88 | |
XM_017023140.1 | XP_016878629.1 | 11 - 12 | 1218 | 88 | |
XM_017023141.1 | XP_016878630.1 | 10 - 11 | 2325 | 88 | |
GenBank mRNA | AJ133798.1 | - | 13 - 14 | 1467 | 88 |
BC035334.1 | - | 11 - 12 | 1181 | 88 | |
BC064577.1 | - | 11 - 12 | 1220 | 88 |