Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NOX3
Entrez Gene ID
50508
Gene Name
NADPH oxidase 3
Gene Aliases
GP91-3, MOX-2
Chromosome Location
Chr.6: 155395368 - 155455903 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015718.2 NP_056533.1 13 - 14 1836 84
GenBank mRNA AF190122.1 - 13 - 14 1836 84

View Full Product Details