The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015718.2 | NP_056533.1 | 9 - 10 | 1249 | 100 |
GenBank mRNA | AF190122.1 | - | 9 - 10 | 1249 | 100 |
AF229177.1 | - | 7 - 8 | 988 | 100 |