The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025132.3 | NP_079408.3 | 4 - 5 | 443 | 85 |
XM_011513724.1 | XP_011512026.1 | 4 - 5 | 353 | 85 | |
XM_011513725.2 | XP_011512027.1 | 3 - 4 | 308 | 85 | |
GenBank mRNA | AK026780.1 | - | 4 - 5 | 318 | 85 |
AK225588.1 | - | 4 - 5 | 318 | 85 | |
AK304161.1 | - | 4 - 5 | 316 | 85 | |
AY029257.1 | - | 4 - 5 | 443 | 85 | |
BC032578.1 | - | 3 - 4 | 322 | 85 |