The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016930.2 | NP_058626.1 | 7 - 8 | 787 | 106 |
XM_011513480.1 | XP_011511782.1 | 8 - 9 | 859 | 106 | |
XM_011513485.2 | XP_011511787.1 | 7 - 8 | 714 | 106 | |
XM_017008297.1 | XP_016863786.1 | 8 - 9 | 1440 | 106 | |
GenBank mRNA | AB028741.1 | - | 7 - 8 | 706 | 106 |
AK310026.1 | - | 7 - 8 | 749 | 106 | |
AL832867.1 | - | 1 - 2 | 53 | 106 | |
AY453396.1 | - | 7 - 8 | 703 | 106 | |
BC014613.2 | - | 7 - 8 | 709 | 106 | |
BT007150.1 | - | 7 - 8 | 703 | 106 | |
BT020135.1 | - | 7 - 8 | 703 | 106 |