Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD37
Entrez Gene ID
951
Gene Name
CD37 molecule
Gene Aliases
GP52-40, TSPAN26
Chromosome Location
Chr.19: 49335314 - 49340608 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040031.1 NP_001035120.1 4 - 5 428 57
NM_001774.2 NP_001765.1 4 - 5 462 57
XM_005259435.3 XP_005259492.1 4 - 5 464 57
XM_005259436.3 XP_005259493.1 4 - 5 460 57
XM_011527542.2 XP_011525844.1 4 - 5 425 57
XM_011527543.2 XP_011525845.1 4 - 5 530 57
XM_011527544.2 XP_011525846.1 4 - 5 499 57
XM_017027513.1 XP_016883002.1 4 - 5 460 57
GenBank mRNA AK301016.1 - 4 - 5 452 57
AK301328.1 - 4 - 5 455 57
AK316348.1 - 4 - 5 352 57
AL833693.1 - 3 - 4 1506 57
BC106752.1 - 4 - 5 379 57
BC110379.1 - 4 - 5 414 57
BT019851.1 - 4 - 5 341 57
CR541877.1 - 4 - 5 341 57
X14046.1 - 4 - 5 404 57

View Full Product Details