The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006724639.3 | XP_006724702.1 | 1 - 2 | 302 | 103 |
GenBank mRNA | AU129897.1 | - | 1 - 2 | 302 | 103 |