Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHMP4C
Entrez Gene ID
92421
Gene Name
charged multivesicular body protein 4C
Gene Aliases
SNF7-3, Shax3, VPS32C
Chromosome Location
Chr.8: 81732453 - 81759515 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152284.3 NP_689497.1 3 - 4 653 63
GenBank mRNA AB120734.1 - 3 - 4 485 63
AK000049.1 - 3 - 4 615 63
AK314873.1 - 3 - 4 652 63
AY329086.1 - 3 - 4 487 63
BC014321.1 - 3 - 4 657 63

View Full Product Details