Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ICMT
Entrez Gene ID
23463
Gene Name
isoprenylcysteine carboxyl methyltransferase
Gene Aliases
HSTE14, MST098, MSTP098, PCCMT, PCMT, PPMT
Chromosome Location
Chr.1: 6221193 - 6235984 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012405.3 NP_036537.1 2 - 3 356 103
GenBank mRNA AF064084.1 - 2 - 3 326 103
AK092890.1 - 3 - 4 297 103
BC028168.1 - 2 - 3 312 103
CR541671.1 - 2 - 3 283 103
CR541711.1 - 2 - 3 283 103

View Full Product Details