Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PKD1L2
Entrez Gene ID
114780
Gene Name
polycystin 1 like 2 (gene/pseudogene)
Gene Aliases
PC1L2
Chromosome Location
Chr.16: 81100879 - 81220370 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278425.1 NP_001265354.1 9 - 10 1443 91
NM_052892.3 NP_443124.3 20 - 21 3349 91
NR_126532.1 20 - 21 3349 91
GenBank mRNA AB067466.1 - 11 - 12 2762 91
AY164483.1 - 20 - 21 3349 91
AY371495.1 - 9 - 10 1405 91

View Full Product Details