The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001288715.1 | NP_001275644.1 | 8 - 9 | 1627 | 78 |
NM_001288716.1 | NP_001275645.1 | 6 - 7 | 889 | 78 | |
NM_001288717.1 | NP_001275646.1 | 8 - 9 | 1254 | 78 | |
NM_001332.3 | NP_001323.1 | 9 - 10 | 1819 | 78 | |
NR_109988.1 | 7 - 8 | 1081 | 78 | ||
XM_005248251.3 | XP_005248308.1 | 9 - 10 | 1843 | 78 | |
XM_005248252.2 | XP_005248309.1 | 9 - 10 | 2009 | 78 | |
XM_005248253.1 | XP_005248310.1 | 8 - 9 | 1534 | 78 | |
XM_011513967.2 | XP_011512269.1 | 8 - 9 | 1923 | 78 | |
XM_017009072.1 | XP_016864561.1 | 7 - 8 | 1093 | 78 | |
XM_017009073.1 | XP_016864562.1 | 7 - 8 | 1268 | 78 | |
XM_017009074.1 | XP_016864563.1 | 7 - 8 | 1090 | 78 | |
XM_017009075.1 | XP_016864564.1 | 6 - 7 | 1142 | 78 | |
XM_017009076.1 | XP_016864565.1 | 6 - 7 | 1153 | 78 | |
GenBank mRNA | AB013805.1 | - | 9 - 10 | 1775 | 78 |
AK127257.1 | - | 3 - 4 | 714 | 78 | |
AK226124.1 | - | 8 - 9 | 1506 | 78 | |
AK294420.1 | - | 5 - 6 | 607 | 78 | |
AK296232.1 | - | 8 - 9 | 1254 | 78 | |
AK299300.1 | - | 6 - 7 | 820 | 78 | |
EU328348.1 | - | 7 - 8 | 1018 | 78 | |
U52351.1 | - | 3 - 4 | 584 | 78 | |
U81004.1 | - | 4 - 5 | 1075 | 78 | |
U96136.1 | - | 9 - 10 | 1629 | 78 |