The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128227.2 | NP_001121699.1 | 5 - 6 | 1187 | 61 |
NM_001190383.1 | NP_001177312.1 | 5 - 6 | 1203 | 61 | |
NM_001190384.1 | NP_001177313.1 | 3 - 4 | 844 | 61 | |
NM_001190388.1 | NP_001177317.1 | 4 - 5 | 981 | 61 | |
NM_005476.5 | NP_005467.1 | 5 - 6 | 1203 | 61 | |
XM_005251334.4 | XP_005251391.1 | 4 - 5 | 1034 | 61 | |
XM_017014167.1 | XP_016869656.1 | 5 - 6 | 1048 | 61 | |
XM_017014168.1 | XP_016869657.1 | 4 - 5 | 1027 | 61 | |
GenBank mRNA | AF051852.1 | - | 4 - 5 | 1032 | 61 |
AF155663.1 | - | 4 - 5 | 982 | 61 | |
AJ238764.1 | - | 4 - 5 | 1023 | 61 | |
AK295562.1 | - | 3 - 4 | 790 | 61 | |
AK296687.1 | - | 4 - 5 | 898 | 61 | |
AK312539.1 | - | 5 - 6 | 1117 | 61 | |
AM697708.1 | - | 5 - 6 | 1075 | 61 | |
AM697709.1 | - | 4 - 5 | 859 | 61 | |
AY531126.1 | - | 4 - 5 | 1023 | 61 | |
AY531127.1 | - | 4 - 5 | 1023 | 61 | |
AY531128.1 | - | 4 - 5 | 1023 | 61 | |
BC121179.2 | - | 5 - 6 | 1111 | 61 | |
EU093084.1 | - | 4 - 5 | 1033 | 61 |