Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EFCAB12
Entrez Gene ID
90288
Gene Name
EF-hand calcium binding domain 12
Gene Aliases
C3orf25
Chromosome Location
Chr.3: 129401321 - 129428651 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207307.2 NP_997190.1 5 - 6 1204 89
XM_011513293.2 XP_011511595.1 5 - 6 3278 89
XM_011513294.2 XP_011511596.1 5 - 6 3276 89
XM_011513295.2 XP_011511597.1 5 - 6 3279 89
XM_011513297.2 XP_011511599.1 5 - 6 3280 89
XM_011513298.1 XP_011511600.1 2 - 3 709 89
XM_011513299.2 XP_011511601.1 2 - 3 903 89
GenBank mRNA AK096099.1 - 3 - 4 630 89
AK127968.1 - 5 - 6 1189 89
AL133011.1 - 3 - 4 4658 89
BC066975.1 - 5 - 6 1204 89

View Full Product Details