Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BBS5
Entrez Gene ID
129880
Gene Name
Bardet-Biedl syndrome 5
Gene Aliases
-
Chromosome Location
Chr.2: 169479496 - 169506655 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152384.2 NP_689597.1 8 - 9 745 137
GenBank mRNA AL834305.1 - 8 - 9 736 137
AY604003.1 - 8 - 9 761 137
BC044593.1 - 8 - 9 742 137
KU178857.1 - 8 - 9 687 137

View Full Product Details