Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BBS5
Entrez Gene ID
129880
Gene Name
Bardet-Biedl syndrome 5
Gene Aliases
-
Chromosome Location
Chr.2: 169479496 - 169506655 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152384.2 NP_689597.1 9 - 10 879 82
GenBank mRNA AK097935.1 - 1 - 2 592 82
AK312680.1 - 8 - 9 796 82
AL834305.1 - 9 - 10 870 82
AY604003.1 - 9 - 10 895 82
AY604004.1 - 8 - 9 832 82
BC044593.1 - 9 - 10 876 82
KU178857.1 - 9 - 10 821 82
KU178858.1 - 8 - 9 758 82

View Full Product Details