The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198443.1 | NP_940845.1 | 1 - 2 | 104 | 85 |
GenBank mRNA | AY358782.1 | - | 1 - 2 | 104 | 85 |
BC100863.2 | - | 1 - 2 | 103 | 85 | |
BC100864.1 | - | 1 - 2 | 103 | 85 | |
BC100865.1 | - | 1 - 2 | 103 | 85 | |
BC100866.2 | - | 1 - 2 | 103 | 85 |