Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TADA2A
Entrez Gene ID
6871
Gene Name
transcriptional adaptor 2A
Gene Aliases
ADA2, ADA2A, KL04P, TADA2L, hADA2
Chromosome Location
Chr.17: 37406877 - 37479725 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166105.2 NP_001159577.2 13 - 14 1188 129
NM_001488.4 NP_001479.4 13 - 14 1396 129
XM_006722044.3 XP_006722107.1 13 - 14 1317 129
XM_011525153.2 XP_011523455.1 12 - 13 1090 129
XM_011525154.2 XP_011523456.1 12 - 13 1309 129
XM_011525155.2 XP_011523457.1 11 - 12 998 129
XM_017024981.1 XP_016880470.1 12 - 13 1124 129
XM_017024982.1 XP_016880471.1 11 - 12 1026 129
XM_017024983.1 XP_016880472.1 13 - 14 1281 129
XM_017024984.1 XP_016880473.1 12 - 13 1105 129
XM_017024985.1 XP_016880474.1 12 - 13 1266 129
XM_017024986.1 XP_016880475.1 12 - 13 1099 129
GenBank mRNA AF069732.1 - 9 - 10 787 129
AK022767.1 - 13 - 14 1399 129
AK096371.1 - 13 - 14 1175 129
AK225918.1 - 13 - 14 1185 129
BC011753.2 - 12 - 13 1135 129
KU178344.1 - 12 - 13 1027 129
KU178345.1 - 11 - 12 938 129

View Full Product Details