Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NUMB
Entrez Gene ID
8650
Gene Name
NUMB, endocytic adaptor protein
Gene Aliases
C14orf41, S171, c14_5527
Chromosome Location
Chr.14: 73275210 - 73458580 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005743.1 NP_001005743.1 11 - 12 1422 80
NM_003744.5 NP_003735.3 10 - 11 1389 80
GenBank mRNA AF171938.1 - 11 - 12 1372 80
AF171940.1 - 10 - 11 1339 80
AK226069.1 - 5 - 6 2958 80
AK315130.1 - 9 - 10 1270 80
BX248073.1 - 10 - 11 1279 80

View Full Product Details