Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NUF2
Entrez Gene ID
83540
Gene Name
NUF2, NDC80 kinetochore complex component
Gene Aliases
CDCA1, CT106, NUF2R
Chromosome Location
Chr.1: 163321933 - 163355764 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031423.3 NP_113611.2 10 - 11 960 86
NM_145697.2 NP_663735.2 10 - 11 1106 86
XM_011510036.1 XP_011508338.1 10 - 11 902 86
GenBank mRNA AB050577.1 - 10 - 11 1021 86
AB050578.1 - 4 - 5 325 86
AF326731.1 - 10 - 11 937 86
AK093348.1 - 10 - 11 1085 86
BC008489.1 - 10 - 11 893 86
BC021171.2 - 4 - 5 375 86

View Full Product Details