Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- NUF2
- Entrez Gene ID
-
83540
- Gene Name
- NUF2, NDC80 kinetochore complex component
- Gene Aliases
-
CDCA1, CT106, NUF2R
- Chromosome Location
-
Chr.1: 163321933 - 163355764 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BI545465.1 |
- |
1 - 2 |
142 |
148 |
View Full Product Details