Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NUF2
Entrez Gene ID
83540
Gene Name
NUF2, NDC80 kinetochore complex component
Gene Aliases
CDCA1, CT106, NUF2R
Chromosome Location
Chr.1: 163321933 - 163355764 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031423.3 NP_113611.2 6 - 7 584 122
NM_145697.2 NP_663735.2 6 - 7 730 122
XM_011510036.1 XP_011508338.1 6 - 7 526 122
GenBank mRNA AB050577.1 - 6 - 7 645 122
AF326731.1 - 6 - 7 561 122
AK093348.1 - 6 - 7 709 122
BC008489.1 - 6 - 7 517 122

View Full Product Details