The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031423.3 | NP_113611.2 | 8 - 9 | 765 | 142 |
NM_145697.2 | NP_663735.2 | 8 - 9 | 911 | 142 | |
XM_011510036.1 | XP_011508338.1 | 8 - 9 | 707 | 142 | |
GenBank mRNA | AB050577.1 | - | 8 - 9 | 826 | 142 |
AB050578.1 | - | 2 - 3 | 130 | 142 | |
AF326731.1 | - | 8 - 9 | 742 | 142 | |
AK093348.1 | - | 8 - 9 | 890 | 142 | |
BC008489.1 | - | 8 - 9 | 698 | 142 | |
BC021171.2 | - | 2 - 3 | 180 | 142 |