Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZMYND10
Entrez Gene ID
51364
Gene Name
zinc finger MYND-type containing 10
Gene Aliases
BLU, CILD22, FLU
Chromosome Location
Chr.3: 50341106 - 50346028 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308379.1 NP_001295308.1 10 - 11 1397 64
NM_015896.3 NP_056980.2 11 - 12 1412 64
XM_005265216.2 XP_005265273.1 10 - 11 1585 64
GenBank mRNA AK058135.1 - 11 - 12 1342 64
AK093455.1 - 3 - 4 1400 64
AK096525.1 - 11 - 12 2520 64
AK223343.1 - 11 - 12 1327 64
AK315123.1 - 10 - 11 1383 64
AL833828.1 - 11 - 12 1341 64
BC033732.1 - 11 - 12 1308 64
U70824.1 - 11 - 12 1366 64
U70880.1 - 10 - 11 1351 64

View Full Product Details