Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAP4
Entrez Gene ID
4134
Gene Name
microtubule associated protein 4
Gene Aliases
-
Chromosome Location
Chr.3: 47850690 - 48089279 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011533710.2 XP_011532012.1 6 - 7 633 82
XM_017006394.1 XP_016861883.1 6 - 7 1082 82
XM_017006401.1 XP_016861890.1 5 - 6 597 82
XM_017006410.1 XP_016861899.1 6 - 7 1097 82
XM_017006411.1 XP_016861900.1 5 - 6 1046 82
XM_017006412.1 XP_016861901.1 5 - 6 1046 82
XM_017006416.1 XP_016861905.1 4 - 5 546 82

View Full Product Details