The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001165966.1 | NP_001159438.1 | 13 - 14 | 1932 | 101 |
NM_031220.3 | NP_112497.2 | 14 - 15 | 2040 | 101 | |
XM_011524015.2 | XP_011522317.1 | 14 - 15 | 1980 | 101 | |
XM_011524016.2 | XP_011522318.1 | 14 - 15 | 1981 | 101 | |
GenBank mRNA | AB209130.1 | - | 8 - 9 | 4403 | 101 |
AF334586.1 | - | 14 - 15 | 2037 | 101 | |
AL389994.1 | - | 13 - 14 | 1763 | 101 | |
BC020484.1 | - | 5 - 6 | 638 | 101 | |
BC128583.1 | - | 14 - 15 | 1977 | 101 | |
BC128584.1 | - | 14 - 15 | 1977 | 101 |