The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001165966.1 | NP_001159438.1 | 3 - 4 | 320 | 70 |
NM_031220.3 | NP_112497.2 | 4 - 5 | 428 | 70 | |
XM_011524015.2 | XP_011522317.1 | 4 - 5 | 368 | 70 | |
XM_011524016.2 | XP_011522318.1 | 4 - 5 | 369 | 70 | |
XM_011524017.2 | XP_011522319.1 | 4 - 5 | 370 | 70 | |
GenBank mRNA | AF334586.1 | - | 4 - 5 | 425 | 70 |
AL389994.1 | - | 3 - 4 | 151 | 70 | |
BC128583.1 | - | 4 - 5 | 365 | 70 | |
BC128584.1 | - | 4 - 5 | 365 | 70 |