Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ALOXE3
Entrez Gene ID
59344
Gene Name
arachidonate lipoxygenase 3
Gene Aliases
ARCI3, E-LOX, eLOX-3, eLOX3
Chromosome Location
Chr.17: 8095900 - 8118916 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001165960.1 NP_001159432.1 1 - 2 219 73
GenBank mRNA AK296416.1 - 1 - 2 120 73
AK299742.1 - 1 - 2 162 73
DC376381.1 - 1 - 2 219 73

View Full Product Details