Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRVI1
Entrez Gene ID
10335
Gene Name
murine retrovirus integration site 1 homolog
Gene Aliases
IRAG, JAW1L
Chromosome Location
Chr.11: 10573091 - 10693988 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098579.2 NP_001092049.2 1 - 2 277 111
NM_001206880.1 NP_001193809.1 2 - 3 611 111
NM_130385.3 NP_569056.4 2 - 3 611 111
GenBank mRNA AK296336.1 - 2 - 3 111
AK300358.1 - 1 - 2 111
AK304702.1 - 2 - 3 111
DA797538.1 - 1 - 2 111
DB167290.1 - 3 - 4 111

View Full Product Details