The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015540.3 | NP_056355.2 | 2 - 3 | 314 | 71 |
XM_005254297.1 | XP_005254354.1 | 2 - 3 | 593 | 71 | |
XM_006720469.1 | XP_006720532.1 | 2 - 3 | 308 | 71 | |
GenBank mRNA | AB037824.1 | - | 2 - 3 | 282 | 71 |
AK290224.1 | - | 2 - 3 | 295 | 71 | |
BC000246.1 | - | 2 - 3 | 296 | 71 | |
DA618150.1 | - | 2 - 3 | 314 | 71 |