The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002248.4 | NP_002239.2 | 3 - 4 | 715 | 93 |
XM_005259905.1 | XP_005259962.1 | 2 - 3 | 673 | 93 | |
XM_005259906.1 | XP_005259963.1 | 2 - 3 | 673 | 93 | |
XM_005259907.2 | XP_005259964.1 | 2 - 3 | 673 | 93 | |
XM_011528004.1 | XP_011526306.1 | 2 - 3 | 673 | 93 | |
XM_011528005.2 | XP_011526307.1 | 3 - 4 | 688 | 93 | |
GenBank mRNA | AB198191.1 | - | 1 - 2 | 459 | 93 |
AK298170.1 | - | 3 - 4 | 661 | 93 | |
BC075037.2 | - | 1 - 2 | 425 | 93 | |
U69883.2 | - | 3 - 4 | 725 | 93 |