Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MASP1
Entrez Gene ID
5648
Gene Name
mannan binding lectin serine peptidase 1
Gene Aliases
3MC1, CRARF, CRARF1, MAP1, MASP, MASP3, MAp44, PRSS5, RaRF
Chromosome Location
Chr.3: 187216085 - 187292220 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001879.5 NP_001870.3 9 - 10 1610 69
NM_139125.3 NP_624302.1 9 - 10 1610 69
NR_033519.1 8 - 9 1378 69
XM_011512989.2 XP_011511291.1 9 - 10 1604 69
XM_011512990.2 XP_011511292.1 9 - 10 1604 69
XM_017006869.1 XP_016862358.1 10 - 11 1688 69
XM_017006870.1 XP_016862359.1 8 - 9 1576 69
XM_017006871.1 XP_016862360.1 9 - 10 1604 69
XM_017006872.1 XP_016862361.1 9 - 10 1463 69
GenBank mRNA AF284421.1 - 9 - 10 1310 69
AK291157.1 - 9 - 10 1446 69
AK291686.1 - 9 - 10 1288 69
AK304334.1 - 8 - 9 1088 69
AL708781.1 - 1 - 2 94 69
BC106945.1 - 9 - 10 1270 69
BC106946.1 - 9 - 10 1270 69
BX647237.1 - 8 - 9 1093 69

View Full Product Details