The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039845.2 | NP_001034934.1 | 3 - 4 | 544 | 90 |
NM_001282940.1 | NP_001269869.1 | 3 - 4 | 544 | 90 | |
NR_104261.1 | 3 - 4 | 544 | 90 | ||
XM_005246315.4 | XP_005246372.1 | 2 - 3 | 365 | 90 | |
XM_005246316.4 | XP_005246373.1 | 3 - 4 | 335 | 90 | |
XM_011510628.2 | XP_011508930.1 | 3 - 4 | 544 | 90 | |
XM_017003373.1 | XP_016858862.1 | 2 - 3 | 213 | 90 | |
XM_017003374.1 | XP_016858863.1 | 2 - 3 | 214 | 90 | |
GenBank mRNA | AK058070.1 | - | 2 - 3 | 191 | 90 |
AK292378.1 | - | 3 - 4 | 542 | 90 | |
AK292386.1 | - | 3 - 4 | 281 | 90 | |
AK302250.1 | - | 3 - 4 | 302 | 90 | |
AK308257.1 | - | 3 - 4 | 321 | 90 | |
AK309144.1 | - | 2 - 3 | 208 | 90 | |
BC033509.1 | - | 3 - 4 | 289 | 90 | |
BC088356.1 | - | 3 - 4 | 298 | 90 | |
BI464613.1 | - | 3 - 4 | 326 | 90 |